Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclindrical Vessels with Verticle Side Arm, CHEMGLASS INC

Supplier: Chemglass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CLS-1406-08 CLS-1406-03 CLS-1406-06 CLS-1404-03 CLS-1406-15 CLS-1404-15 CLS-1404-08 CLS-1406-36 CLS-1404-06
89166-838EA 1559.38 USD InStock
89166-838 89166-850 89166-840 89166-852 89166-842 89166-844 89166-846 89166-836 89166-848
Cyclindrical Vessels with Verticle Side Arm, CHEMGLASS INC
Reaction Vessels Reaction Vessels, Cylindrical Shape
Bioreactor vessels are ideal for cell cultivation and fermentation.

  • Four or six vertical side-arms with GL 45 opening
  • Without baffles
  • Center joint: 100 mm

The 3 and 6 L vessels have a low profile, it reduced vessel height maximizes incubator space.

Delivery information: Supplied with four or six solid GL 45 caps and pouring rings.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty