Would you like to log in again?
Anaerobic Hungate Culture Tubes, GPI Screw Threads, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CLS-4208-01
89167-170PK 121.57 USD InStock
89167-170
Anaerobic Hungate Culture Tubes, GPI Screw Threads, Chemglass
Tubes Culture Tubes
Anaerobic hungate culture tubes are designed to maintain anaerobic culture conditions.
Flange style stoppers are made from butyl rubber. Complete item consists of a disposable glass tube, butyl stopper, and screw cap with a 9 mm opening.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty