Would you like to log in again?
Filter Funnels, Disposable, Polyethylene Frits, Chemglass
Supplier: Chemglass
OP-6602-10 OP-6602-14 OP-6602-18 OP-6602-12
89167-224CS 344.8 USD InStock
89167-224 89167-222 89167-220 30617-998
Filter Funnels, Disposable, Polyethylene Frits, Chemglass
Funnels Filtration Funnels Filter Funnels
This unique disposable filter funnel system for solid-liquid separation can be used in chemistry and biological laboratories.
The system consists of a polypropylene filter funnel with a 10 micron polyethylene fritted disc, style glass vacuum adapter, and a flask or vial
The disposable filter funnels have a 80mm long stem so that the glass vacuum adapter is not contaminated during the filtration process.
The vacuum adapters are specially designed so that the vacuum can be controlled using a Teflon stopcock to prevent the filtrates from being sucked into the vacuum line.
Packaging: Bulk packed in a zip lock bag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty