Would you like to log in again?
E.Z.N.A. FFPE RNA Kit, Omega Bio-tek
Supplier: Omega Bio-Tek
E.Z.N.A®
R6954-01 R6954-00 R6954-02 R6954-00 R6954-01
89167-338EA 381.26 USD InStock
CA89167-338 89167-338 CA89167-336 89167-336 CA89167-340
E.Z.N.A. FFPE RNA Kit, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® FFPE RNA Kit is specially designed for isolation of total RNA from formalin-fixed, paraffin-embedded tissue sections
This method can also be used for the preparation of total RNA from fibrous samples such as heart and muscles tissue. The sample is first processed in special buffer by heating to deactivate RNase’s and deparaffinization. Then the lysate is treated with Proteinase K to digest proteins. The cleared lysate is then applied to a HiBind RNA column to capture RNA. After two quick wash steps, purified RNA is eluted with water. While E.Z.N.A.® FFPE RNA Kit is optimized to minimize the effect of the formaldehyde modification, it is not recommended to use the RNA purified from this kit for downstream applications that requires full length RNA.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty