Would you like to log in again?
Protocol™ Parasitology Vials, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Protocol™
23-005-28 23-005-47 23-005-26 23-005-44 23-005-31 23-005-27 23-005-43 23-005-32 23-005-45 23-005-37 23-005-46 23-005-33 23-005-41 23-005-30 23-005-39
89167-490PK 80.31 USD InStock
89167-490 89167-508 89167-506 89167-504 89167-502 89167-488 89167-510 89167-500 89167-486 89167-498 89167-484 89167-496 89167-482 89167-494 89167-492
Protocol™ Parasitology Vials, Thermo Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Faeces Sample Collection
For the collection, transport, and examination of fecal specimens
Sodium acetate-acetic acid-formalin (SAF) is ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, and cocciadian oocysts and microsporidian spores. Slides fixed with SAF may be stained using trichrome or hematoxylin. 10% neutral buffered formalin is an all purpose fixative appropriate for helminth eggs and larvae and for protozoan cysts. The buffering system helps to maintain organism morphology. Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae, and protozoan cysts.
C&S media and Cary Blair media facilitate the collection and transport of enteric bacterial pathogens, while maintaining the bacterial population for optimum recovery for up to 96 hours. C&S medium: Agar, sodium thioglycolate, Na2HPO4, calcium chloride, phenol red. Cary Blair medium: Agar, DI water, KH2PO4, NA2HPO4, phenol red, sodium chloride, sodium thioglycolate.
Modified (Cu) PVA fixative acts as a preservative and fixative ideally suited for protozoan trophozoites. Slides fixed with Modified PVA may be stained with trichrome or hematoxylin.
Zn-PVA is used as a preservative and fixative for protozoan trophozoites. Proper use assures the parasitologist that diagnostic stages of intestinal parasites, if present, will be preserved. Slides fixed Zn-PVA may be stained using trichrome or hematoxylin.
A full line of mercury-free fixatives and preservatives are available in 15mL single vials, or double- and triple-vial kits to meet individual laboratory needs
Ordering information: Single-vial kits contain 20 vials. Double- and triple-vial kits contain ten vials of each type.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty