Would you like to log in again?
Axygen® GEN3 Multi-Barrier Filter Tips, Sterile, Corning
Supplier: Corning
Axygen®
GEN3-20LRS GEN3-10LRS GEN3-200LRS GEN3-1000-L-R-S
89167-688CS 2706.16 USD InStock
89167-688 89167-686 89167-692 89167-690
Axygen® GEN3 Multi-Barrier Filter Tips, Sterile, Corning
Pipette Tips
The new Axygen brand GEN3 multi-barrier filter pipette tips offer the ultimate in protection against cross contamination through tri-filter technology.
  • Offered in racks and sterile
  • Certified human gDNA and PCR inhibition free, DNase-/RNase-free, and nonpyrogenic
  • Prevents liquid or aerosol from passing through and will lock on contact
GEN3 tips effectively separate liquid from the self-sealing barrier using an additional protective layer of conventional polyethylene filter material. In addition to the self-sealing properties, GEN3 multi-barrier pipette tips feature the exclusive Maxymum Recovery® technology, which reduces the amount of reagent or sample adhering to the pipette tips.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty