Would you like to log in again?
Ionic Detergent Solutions, G-Biosciences
Supplier: G-Biosciences
DG092 DG094 DG094 786-017 786-016 DG093 DG093 DG091 DG095 DG090 DG092 DG091 DG095 786-016 DG090 786-017
82023-052EA 110.25 USD InStock
82023-052 CA82023-056 82023-058 CA89167-708 82023-048 CA82023-058 CA89167-706 CA82023-052 89167-708 82023-056 89167-706 95029-300 CA95029-300 95029-302 CA95029-302 CA82023-048
Ionic Detergent Solutions, G-Biosciences
Cleaning Agents Detergents and Surface Cleansers
Ionic detergents have a hydrophilic head group that is charged and can be either negatively (anionic) or positively (cationic) charged
G-Biosciences Ionic Detergent Solutions include: SDS, CTAB, and Deoxycholate.
Ionic detergents are used for the complete disruption of cellular structures and denaturation of proteins for separation during gel electrophoresis. Ionic detergents bind with protein molecules, masking their native charge and rendering the protein molecules the overall charge of the ionic detergent.
Ordering information: Accessories not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty