Would you like to log in again?
Protein Concentrator Solution, G-Biosciences
Supplier: G-Biosciences
786-143
89167-726EA 166.45 USD InStock
89167-726 CA89167-726
Protein Concentrator Solution, G-Biosciences
Protein Purification Protein Purification Reagents
Concentrator Solution is a novel liquid polymer for the rapid concentration of dilute protein solutions with zero loss, using dialysis
The water will be rapidly removed through the dialysis membrane, which also retains your protein of interest and prevents the high molecular weight liquid polymer entering your solution. Once the desired volume of your solution is achieved, quickly rinse the excess concentrator solution from the dialysis bag/ membrane and recover your sample.
Concentrator Solution is also supplied with our Tube-O-DIALYZER™ as Tube-O-CONCENTRATOR™. Use molecular weight cut offs (MWCO) of 30,000 daltons or less to avoid cross contamination.
Simply transfer your dilute protein solution to a dialysis bag or dialysis device, such as our patented Tube-O-DIALYZER™ and dialyze against the concentrator solution.
Ordering information: Accessories not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty