Would you like to log in again?
Fluorescent β-Galactosidase Assay, G-Biosciences
Supplier: G-Biosciences
786-654 786-654
89167-842EA 779.13 USD InStock
CA89167-842 89167-842
Fluorescent β-Galactosidase Assay, G-Biosciences
Assays Enzyme Assays
G-Biosciences' Fluorescent β-Galactosidase Assay (MUG) is a highly sensitive, fluorescent assay for determining the β-galactosidase activity in the lysates of cells transfected with a β-galactosidase expression construct.
The study of the lac operon has played an important role in understanding the control of gene expression in bacteria. In prokaryotes, gene expression is controlled primarily at the level of transcription. For eukaryotes, the promoter activity can be analyzed by using fusion genes containing the promoter of interest attached to the bacterial β-galactosidase gene and be assayed by measuring β-galactosidase activity. Because β-galactosidase has a high turnover rate and is absent in mammalian cells it serves as a very useful and sensitive reporting tool for gene expression.
Esters of the fluorescent compound, 4-methylumbelliferone (4-MU), provide a sensitive, quantitative assay for β-galactosidase. 4-methylumbelliferyl-β-D-galactopyranoside (4-MUG) is a substrate of β-galactosidase that does not fluoresce until cleaved by the enzyme to generate the fluorophore 4-methylumbelliferone. The assay can be used with extracts from different expression systems including mammalian, insect cells, yeast, and bacteria.
G-Biosciences' Fluorescent β-Galactosidase Assay (MUG) provides a 96 well assay format for galactosidase activity that is suitable for high throughput applications. The production of the fluorphore is monitored at an emission/excitation wavelength of 365/460nm. This kit is designed for 500 micro-well assays.
Ordering information: Accessories available
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty