Would you like to log in again?
Protein-Free™ Blocking Buffers, G-Biosciences
Supplier: G-Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
786-662 786-665 786-664 786-663
89167-858EA 164.52 USD InStock
89167-858 CA89167-864 89167-864 CA89167-862 89167-862 CA89167-860 89167-860 CA89167-858
Protein-Free™ Blocking Buffers, G-Biosciences
Buffers Electrophoresis Buffers Blocking Buffers
G-Biosciences' Protein-Free™ Blocking Buffer does not contain protein; it is a proprietary formation of non-protein agents that eliminates non-specific binding sites in ELISA, blotting, immunohistochemistry and other applications
G-Biosciences' Protein-Free™ Blocking Buffer eliminates any concern associated with regulatory compliance issues where use of animal source components are restricted. Furthermore, Protein-Free™ Blocking Buffer is compatible with antibodies and avidin/biotin based systems and results in high signal to background ratios.
G-Biosciences' Protein-Fre™ Blocking Buffer is offered in four convenient ready-to-use formats, with or without detergent: supplied either in TBS buffer (Tris-buffered saline at pH 7.5) or PBS buffer (phosphate-buffered saline at pH 7.5), as well as in separate formulations that contain G-Biosciences' detergent Tween®-20 for improving blocking efficiencies.
The absence of protein from the Protein-Free™ Blocking Buffer eliminates problems associated with traditional protein based blockers, such as cross-reactivity and interference from glycosylated proteins.
Ordering information: Accessories available.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty