Would you like to log in again?
Immobilized Trypsin
Supplier: G-Biosciences
786-792 786-792
89168-040EA 166.65 USD InStock
89168-040 CA89168-040
Immobilized Trypsin
Enzymes
Immobilized Trypsin is TPCK Treated Trypsin immobilized on 4% agarose that eliminates the contamination of protein digests by the trypsin
Trypsin is a serine endopeptidase that specifically cleaves peptide bonds on the carboxy side of s-aminoethyl cysteine, arginine and lysine residues and typically there is little or no cleavage at arginyl-proline and lysyl-proline bonds. The distribution of these residues in proteins allows trypsin digestion to produce peptides that are readily identified by mass spectrometry. Native trypsin is prone to autolysis that results in pseudotrypsin, which exhibits a broader proteolytic specificity (a chymotrypsin like activity) and trypsin fragments that interfere with sequence analysis.
The Trypsin is TPCK treated to inactive the interfering chymotrypsin activity and the resulting protein is affinity purified. Immobilzed Trypsin is supplied as a 50% slurry containing glycerol and sodium azide as a preservative.
The immobilized trypsin is readily removed by separating the agarose from the digestion solution.
Ordering information: Accessories not included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty