Would you like to log in again?
Sulfhydryl Coupling Resin for Antibody Purification, G-Biosciences
Supplier: G-Biosciences
786-794 786-806 786-795 786-806 786-796 786-804 786-805 786-794 786-804 786-805 786-796 786-795
89168-048EA 3928.65 USD InStock
CA89168-046 89168-048 CA89168-048 71003-276 CA71003-274 89168-044 CA89168-044 CA71003-276 89168-046 71003-272 CA71003-272 71003-274
Sulfhydryl Coupling Resin for Antibody Purification, G-Biosciences
Protein Purification Protein Purification Resins
G-Biosciences' Sulfhydryl Coupling Resin is designed for the simple and efficient coupling of peptides and proteins to a solid 6% agarose support through free sulfhydryl groups (-SH)
G-Biosciences' Sulfhydryl Immobilization Kit for Proteins is a complete kit designed for the simple and efficient coupling of proteins to a solid agarose support. Proteins, including antibodies, must have free sulfhydryls for immobilization to the resin. A mild reducing agent, 2-Mercaptoethylamine, is supplied to reduce the hinge region disulfide bonds of antibodies, while preserving the functionally crucial disulfide bonds between the heavy and light chains. The resulting columns can be used to study protein-protein interactions or for purification, via affinity chromatography. The columns, depending on the stability of the immobilized molecule, can be used several times without significant loss of activity.
G-Biosciences' Sulfhydryl Immobilization Kit for Peptides is designed for the simple and efficient coupling of sulfhydryl-containing peptides to a solid agarose support. Peptides must have free sulfhydryls for immobilization to the resin. The supplied Protein-S-S-Reductant™ reducing agent efficiently reduces disulfide bonds and does not interfere with the iodoacetyl coupling reaction. Protein-S-S-Reductant™ offers the advantage that it does not require removal before peptide immobilization. The resulting columns can be used for the purification of antibodies that have been raised against the specific peptide. The columns, depending on peptide stability, can be used several times.
The iodoacetyl groups of the Sulfhydryl Coupling Resin specifically react with free sulfhydryls to form covalent, permanent thioether bonds. The long spacer arm reduces steric hindrance and ensures greater binding of proteins and antibodies during affinity purification. The Sulfhydryl Coupling Resin is available as a resin slurry or prealiquoted as five 2ml spin column format.
Ordering information: Accessories included. Additional supplies available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty