Would you like to log in again?
Pearl™ Monoclonal IgG Purification, G-Biosciences
Supplier: G-Biosciences
786-802 786-802
89168-060EA 2008.62 USD InStock
89168-060 CA89168-060
Pearl™ Monoclonal IgG Purification, G-Biosciences
Protein Purification Protein Purification Kits
G-Biosciences' Pearl™ Monoclonal IgG Purification kit allows for the rapid purification of antibodies from cell culture supernatant and ascites fluid
G-Biosciences' Pearl™ Monoclonal IgG Purification kit can be used to purify antibodies direct from cell culture supernatant with less than 10% FBS or can be used with ascites fluid after treatment with the supplied Ascites PreTreat.
G-Biosciences' Pearl™ Monoclonal IgG Purification kit can purify IgG from ~1L cell culture supernatant or 200 mL ascites fluid.
The Pearl™ IgG Purification Resin binds the high abundant, non-IgG proteins (i.e. albumin) and allows the IgG molecules to pass through in a physiological buffer. The IgG molecules can be stored or used in downstream applications without further clean-up, such as ammonium sulfate precipitation.
Ordering information: Accessories included. Additional supplies available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty