Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Motic BA310 Upright Compound Microscope
Supplier: Motic
1101001700981 1101001702921 1101001901581 1101001903141 1101001901581 1101001701472 1101001702921 1101001402441 1101001903141 1101001701332 1101001701472 1101001701431 1101001701041 1101001402441 1100100401384
89130-142EA 2400.08 USD InStock
89130-142 CA19000-530 CA89168-452 CA89168-474 89168-450 CA89168-470 89168-452 89168-474 CA89168-450 89168-470 CA19000-528 CA19000-522 19000-522 CA19000-526 CA19000-524
Motic BA310 Upright Compound Microscope
Microscopes
The Motic BA310 is designed for the rigors of daily routine work
The Motic BA310 features a Siedentopf style head inclined at 30° and featuring full 360° rotation for comfortable viewing over extended periods. Also included are Widefield high eyepoint eyepieces, CCIS EF-N Plan Achromat objectives (4x, 10x, 40x S, and 100x S-oil), coaxial coarse and fine focusing adjustment, a built-in low position coaxial mechanical stage (right-hand control), and a focusable Abbe condenser with iris diaphragm and slot. All inclusions help ensure high quality, reproducible images. Additional optional contrast methods such as phase contrast, polarization, and darkfield (as well as discussion/teaching devices) ensure that this model will offer long term functionality to all user levels.
The requirements of any routine microscope demand careful attention to detail on every microscope detail. The BA310 provides just this, with a powerful adjustable light source to meet both delicate and detailed lighting requirements for all sample types, and full Koehler illumination to help in highlighting even the weakest stained specimens. The CCIS EF-N Plan Achromat objectives provide optimal image contrast through engineered multi-layer lens coatings to give superb image clarity and flatness of field.
Ordering information: Microscope is supplied with all accessories listed above. Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty