Would you like to log in again?
Motic BA310 Elite Upright Compound Microscope
Supplier: Motic
1100100402641 1100100402631 1101001100401 1100100402631 1100100402641 1101001100401
10768-196EA 2274.85 USD InStock
10768-196 CA10769-650 CA89168-482 CA10769-646 89168-482 10768-192
Motic BA310 Elite Upright Compound Microscope
Microscopes
Motic’s BA310 Elite Series is built for the performance demands of colleges, universities, and medical school markets, setting a new price-performance standard for optical quality
The BA310 Elite model introduces superior improvements in both optical and mechanical performance. The rackless stage concept without a prominent gear rack permits even more convenient x/y movement and effortless positioning of the stage without gear rack interference. Ergonomic drop-down coaxial controls enable convenient 'eyes down' operation for uninterrupted sample viewing. The included CCIS EC Plan Achromatic lenses are multilayer-coated for improved color fidelity and image enhancement, giving superior image reproduction.
The improved BA310 Elite series introduces innovative features like second-generation EC Plan optics, a new rackless stage movement, and an advanced modular illumination design offering halogen/LED interchangeability. The BA310 Elite series brings versatility and function to a wide range of education and professional applications.
Ordering information: Microscope is customizable with various accessories (sold separately). For all available options, please contact your local Avantor sales representative. All Motic products come with a one-year electrical and five-year mechanical warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty