Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ERGOMAT® Softline® Rubber Mats, ERGOMAT

Supplier: Ergomat

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Ergomat®
SL0420-BLU SL0203-BLU SL0305-BLU SL0305-BLU SL0310-BLU SL0218-BLU SL0210-BLU SL0204-BLU SL0306-BLU SL0406-BLU SL0220-BLU SL0314-BLU SL0205-BLU SL0215-BLU SL0415-BLU SL0215-BLU SL0211-BLU SL0203-BLU SL0411-BLU SL0418-BLU SL0216-BLU SL0217-BLU SL0308-BLU SL0220-BLU SL0212-BLU SL0303-BLU SL0419-BLU SL0318-BLU SL0408-BLU SL0415-BLU SL0416-BLU SL0206-BLU SL0406-BLU SL0311-BLU SL0213-BLU SL0307-BLU SL0304-BLU SL0310-BLU SL0420-BLU SL0317-BLU SL0216-BLU SL0205-BLU SL0414-BLU SL0309-BLU SL0411-BLU SL0206-BLU SL0313-BLU SL0309-BLU SL0312-BLU SL0316-BLU SL0211-BLU SL0210-BLU SL0208-BLU SL0207-BLU SL0412-BLU SL0209-BLU SL0311-BLU SL0304-BLU SL0203-GRA SL0306-BLU SL0315-BLU SL0412-BLU SL0312-BLU SL0209-BLU SL0405-BLU SL0410-BLU SL0405-BLU SL0317-BLU SL0219-BLU SL0315-BLU SL0304-GRA SL0218-BLU SL0404-BLU SL0203-GRA SL0413-BLU SL0408-BLU SL0318-BLU SL0314-BLU SL0214-BLU SL0214-BLU SL0404-BLU SL0407-BLU SL0413-BLU SL0414-BLU SL0219-BLU SL0207-BLU SL0319-BLU SL0313-BLU SL0204-BLU SL0410-BLU SL0409-BLU SL0416-BLU SL0217-BLU SL0320-BLU SL0417-BLU SL0417-BLU SL0319-BLU SL0212-BLU SL0308-BLU SL0307-BLU SL0407-BLU SL0320-BLU SL0213-BLU SL0409-BLU SL0418-BLU SL0303-BLU SL0304-GRA SL0419-BLU SL0208-BLU SL0316-BLU
89169-258EA 1974.75 USD InStock
CA22231-168 CA22231-166 CA22231-164 CA22231-162 CA89169-300 89169-258 CA89169-266 89169-256 CA89169-264 CA89169-304 CA89169-302 CA89169-268 89169-250 89169-254 CA89169-262 89169-252 CA89169-260 22231-168 CA89169-306 22231-166 22231-164 22231-162 CA22231-172 CA89169-278 89169-268 89169-302 89169-266 CA89169-276 89169-300 89169-306 89169-304 89169-260 CA89169-270 89169-264 CA89169-274 89169-262 CA89169-272 CA89169-288 22231-172 89169-278 CA89169-286 CA89169-280 89169-272 89169-270 89169-276 CA89169-284 CA89169-282 89169-274 CA89169-208 CA89169-212 89169-288 CA89169-298 CA89169-210 CA89169-216 CA89169-214 89169-282 CA89169-292 CA89169-290 89169-280 CA89169-296 89169-286 CA89169-294 89169-284 89169-290 CA89169-218 89169-208 89169-214 CA89169-222 CA89169-220 89169-212 89169-218 CA89169-226 CA89169-224 89169-216 89169-294 89169-292 89169-210 89169-298 89169-296 CA89169-228 CA89169-234 89169-224 89169-222 CA89169-232 CA89169-238 89169-228 CA89169-236 89169-226 CA89169-230 89169-220 CA89169-244 89169-236 CA89169-242 89169-234 CA89169-248 89169-238 CA89169-246 89169-232 CA89169-240 89169-230 CA89169-256 89169-246 89169-244 CA89169-254 CA89169-258 89169-248 89169-242 CA89169-252 CA89169-250 89169-240
ERGOMAT® Softline® Rubber Mats, ERGOMAT
Floor Mats Traction and Anti-Fatigue Floor Mats
Pure nitrile rubber mats are ideal for areas exposed to significant moisture and slip hazards

  • These mats are latex-free
  • Mats are autoclavable at 121 °C (250 °F)
  • May be easily cleaned using chemical, steam, or high pressure systems to prevent bacterial accumulation

These nonslip mats promote muscle stimulation, even while standing, which increases circulation and reduces fatigue and long-term injury. Mats are chemical-resistant, heat- and abrasion-resistant, nonporous, nonabsorbent, and formulated to withstand animal grease, gasoline, oil, and organic solvents.

Mats are typically used in metal and welding operations, heavy manufacturing, and commercial kitchens.

Certifications: Mats are suitable for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanrooms.

Ordering information: Mats come with a manufacturer’s three-year warranty. Mats are available in any size or shape; contact your VWR sales representative for more information.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty