The ERGOMAT® Hygiene is a cone profile ergonomic mat well suited for use in critical environments requiring daily caustic sanitation
- Use in hospitals and clinics, laboratories, food processing facilities, commercial kitchens
- Available in 4 colors (blue, gray, red, and green) to help eliminate the potential for cross contamination issues
- Mats can be safely autoclaved (2’ x 3’ size only)
- Mats come with manufacturer’s three-year warranty
Mats are 11 mm (7/16") thick and 91.4 cm (36") wide, with beveled edges on all sides to prevent curling and eliminate trip hazards. These nonslip rubber mats promote occupational safety and health and deliver ergonomic benefits in the most demanding environments.
Mats have a broad range of pH resistance (2 to 13) and tolerate a variety of chemical exposures, except oils.
Certifications: Mats are USDA approved, meet CIFA standards for food processing, and are ISO Class 5 cleanroom rated.