Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ergomat® Hygiene Cleanroom Anti-Fatigue Food and Medical Floor Mats, Ergomat

Supplier: ERGOMAT INC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

ERGOMAT®
EH0218-RED EH0417-RED EH0416-RED EH0411-RED EH0308-RED EH0407-RED EH0416-RED EH0217-RED EH0211-RED EH0418-RED EH0318-RED EH0404-RED EH0206-RED EH0220-RED EH0409-RED EH0210-RED EH0307-RED EH0203-RED EH0206-RED EH0306-RED EH0303-RED EH0215-RED EH0305-RED EH0316-RED EH0310-RED EH0218-RED EH0204-RED EH0216-RED EH0311-RED EH0311-RED EH0214-RED EH0205-RED EH0410-RED EH0217-RED EH0405-RED EH0214-RED EH0414-RED EH0304-RED EH0313-RED EH0306-RED EH0220-RED EH0413-RED EH0410-RED EH0203-RED EH0219-RED EH0404-RED EH0207-RED EH0319-RED EH0305-RED EH0216-RED EH0417-RED EH0414-RED EH0407-RED EH0309-RED EH0406-RED EH0319-RED EH0315-RED EH0312-RED EH0317-RED EH0309-RED EH0213-RED EH0219-RED EH0318-RED EH0207-RED EH0320-RED EH0215-RED EH0312-RED EH0314-RED EH0213-RED EH0308-RED EH0310-RED EH0409-RED EH0209-RED EH0315-RED EH0418-RED EH0415-RED EH0307-RED EH0208-RED EH0320-RED EH0406-RED EH0419-RED EH0210-RED EH0408-RED EH0314-RED EH0204-RED EH0420-RED EH0303-RED EH0304-RED EH0408-RED EH0313-RED EH0413-RED EH0405-RED EH0415-RED EH0208-RED EH0316-RED EH0211-RED EH0412-RED EH0419-RED EH0209-RED EH0212-RED EH0205-RED EH0317-RED EH0412-RED EH0420-RED EH0212-RED EH0411-RED
89169-408EA 394.68 USD InStock
CA89169-418 89169-408 22231-158 CA89169-416 22231-156 CA22231-160 89169-412 CA89169-420 89169-498 89169-410 CA89169-424 89169-416 89169-414 CA89169-422 89169-492 89169-490 89169-496 89169-494 CA89169-428 CA89169-426 89169-418 CA89169-432 22231-160 89169-422 89169-420 CA89169-430 89169-426 CA89169-436 CA89169-434 89169-424 CA89169-438 89169-428 89169-434 CA89169-442 89169-432 CA89169-440 89169-438 CA89169-446 CA89169-444 89169-436 89169-430 CA89169-448 CA89169-454 89169-444 89169-442 CA89169-452 89169-448 CA89169-458 89169-446 CA89169-456 CA89169-450 89169-440 CA89169-464 89169-456 89169-454 CA89169-462 CA89169-502 CA89169-468 CA89169-500 89169-458 CA89169-466 89169-452 CA89169-460 89169-450 CA89169-506 CA89169-504 89169-466 CA89169-476 89169-500 CA89169-474 89169-464 89169-504 CA89169-478 89169-468 89169-502 89169-462 CA89169-472 CA89169-470 89169-460 89169-506 CA89169-486 89169-478 89169-476 CA89169-484 CA89169-488 89169-470 89169-474 CA89169-482 CA89169-480 89169-472 CA22231-158 CA22231-156 CA89169-408 89169-488 CA89169-498 CA89169-410 89169-486 CA89169-496 CA89169-414 CA89169-412 89169-480 CA89169-490 89169-484 CA89169-494 CA89169-492 89169-482
Ergomat® Hygiene Cleanroom Anti-Fatigue Food and Medical Floor Mats, Ergomat
Floor Mats Traction and Anti-Fatigue Floor Mats
Hygiene Food and Medical Mat has a broad chemical and pH resistance profile making it perfect for use in areas requiring frequent and caustic sanitation like medical and food processing applications.

  • Made of Virgin EPDM rubber
  • Can be pressure washed to 1500 psi
  • Broad pH resistance from 2 to 13
  • Service temperature range −40 to 120 °C

In situations with excessive fluid volume, or the need to promote air flow, Hygiene can be die-cut to accommodate these requirements,
Light Weight and Easy to clean has made this mat a favorite amongst medical professionals as well as in the food industry.

Certifications: ISO Cleanroom Class 5 (Class 100) rated and USDA Certified.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty