Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ERGOMAT® Hygiene Rubber Mats, ERGOMAT

Supplier: Ergomat

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

ERGOMAT® Ergomat®
EH0204-GRN EH0314-GRN EH0316-GRN EH0405-GRN EH0204-GRN EH0410-GRN EH0318-GRN EH0216-GRN EH0316-GRN EH0210-GRN EH0418-GRN EH0404-GRN EH0419-GRN EH0406-GRN EH0310-GRN EH0405-GRN EH0203-GRN EH0408-GRN EH0414-GRN EH0208-GRN EH0414-GRN EH0216-GRN EH0416-GRN EH0306-GRN EH0206-GRN EH0419-GRN EH0211-GRN EH0318-GRN EH0320-GRN EH0315-GRN EH0214-GRN EH0215-GRN EH0420-GRN EH0413-GRN EH0215-GRN EH0309-GRN EH0207-GRN EH0320-GRN EH0304-GRN EH0217-GRN EH0409-GRN EH0317-GRN EH0418-GRN EH0314-GRN EH0212-GRN EH0218-GRN EH0305-GRN EH0410-GRN EH0218-GRN EH0308-GRN EH0220-GRN EH0319-GRN EH0319-GRN EH0411-GRN EH0307-GRN EH0412-GRN EH0412-GRN EH0407-GRN EH0205-GRN EH0207-GRN EH0309-GRN EH0208-GRN EH0313-GRN EH0310-GRN EH0311-GRN EH0203-GRN EH0304-GRN EH0420-GRN EH0312-GRN EH0404-GRN EH0217-GRN EH0303-GRN EH0305-GRN EH0209-GRN EH0209-GRN EH0409-GRN EH0406-GRN EH0311-GRN EH0313-GRN EH0211-GRN EH0408-GRN EH0315-GRN EH0213-GRN EH0415-GRN EH0214-GRN EH0417-GRN EH0308-GRN EH0213-GRN EH0210-GRN EH0312-GRN EH0220-GRN EH0219-GRN EH0212-GRN EH0219-GRN EH0306-GRN EH0303-GRN EH0417-GRN EH0205-GRN EH0307-GRN EH0317-GRN EH0411-GRN EH0416-GRN EH0413-GRN EH0415-GRN EH0206-GRN EH0407-GRN
89169-528EA 1434.33 USD InStock
CA89169-538 89169-528 CA89169-536 CA89169-542 89169-532 89169-530 CA89169-540 CA89169-546 89169-536 CA89169-544 89169-534 CA89169-548 89169-538 CA89169-552 89169-544 CA89169-550 89169-542 89169-548 CA89169-556 CA89169-554 89169-546 89169-540 CA89169-558 CA89169-564 89169-554 89169-552 CA89169-562 CA89169-568 89169-558 CA89169-600 CA89169-566 89169-556 89169-550 CA89169-560 CA89169-604 CA89169-602 CA89169-606 CA89169-574 89169-566 89169-564 CA89169-572 89169-602 CA89169-578 89169-600 89169-568 CA89169-576 CA89169-570 89169-562 89169-560 89169-606 89169-604 89169-576 CA89169-586 89169-574 CA89169-584 89169-578 CA89169-588 CA89169-582 89169-572 89169-570 CA89169-580 CA22231-138 CA89169-508 89169-588 CA89169-596 CA89169-594 89169-586 CA89169-512 CA89169-598 CA89169-510 89169-580 CA89169-592 89169-584 CA89169-590 89169-582 22231-138 89169-508 CA89169-516 CA89169-514 CA22231-142 CA22231-140 CA89169-518 CA89169-520 89169-510 89169-598 89169-596 89169-514 CA89169-524 89169-512 CA89169-522 89169-590 89169-594 89169-592 CA89169-528 89169-518 89169-516 CA89169-526 22231-142 22231-140 CA89169-530 89169-522 89169-520 89169-526 CA89169-534 CA89169-532 89169-524
ERGOMAT® Hygiene Rubber Mats, ERGOMAT
Floor Mats Traction and Anti-Fatigue Floor Mats
The ERGOMAT® Hygiene is a cone profile ergonomic mat that is well suited for use in critical environments requiring daily caustic sanitation.

  • Hygiene mats have a broad range of pH resistance (2 to 13), and tolerate a variety of chemical exposures (except oils)
  • Use in hospitals and clinics, laboratories, food processing facilities, and commercial kitchens
  • Available in 4 colors (blue, gray, red, and green) to help eliminate the potential for cross contamination issues
  • They can also be safely autoclaved (2’ x 3’ size only)

Mats are 11 mm (7/16") thick and 91.4 cm (36") wide, with beveled edges on all sides to prevent curling and eliminate trip hazards. These nonslip rubber mats promote occupational safety and health and deliver ergonomic benefits in the most demanding environments.

Certifications: They are USDA approved, meet CIFA standards for food processing, and are ISO Class 5 cleanroom rated.

Ordering information: Mats come with manufacturer’s three-year warranty.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty