Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ergomat® Nitril and Nitril Smooth Cleanroom Anti-Fatigue Floor Mats, Ergomat

Supplier: Ergomat

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

ERGOMAT® Ergomat®
ENS0415 ENS0210 EN0404 ENS0208 ENS0214 ENS0407 ENS0411 ENS0409 EN0317 ENS0406 ENS0308 ENS0207 ENSO304 EN0406 EN0319 EN0211 EN0410 EN0205 EN0305 EN0217 EN0316 EN0303 EN0208 ENS0316 EN0419 EN0318 ENS0211 EN0417 ENS0220 EN0209 EN0319 EN0212 ENS0307 ENS0404 EN0216 ENS0408 ENS0206 ENS0309 EN0414 ENS0307 EN0420 ENS0318 EN0320 EN0210 ENS0305 EN0213 ENS0217 EN0310 EN0211 EN0206 EN0409 ENS0318 EN0307 EN0308 EN0416 ENS0412 EN0209 ENS0217 ENSO206 ENS0209 ENS0216 EN0415 ENS0211 EN0212 ENS0405 EN0215 ENS0212 EN0205 ENS0208 ENS0404 EN0214 ENS0216 ENS0420 EN0309 EN0406 EN0311 ENS0411 ENS0218 ENS0414 ENS0415 EN0410 EN0317 ENS0310 ENS0207 EN0409 EN0304 ENS0316 ENS0413 ENS0419 EN0207 EN0314 EN0217 ENS0420 EN0206 EN0210 EN0411 EN0414 EN0315 EN0413 ENS0311 ENS0306 EN0405 EN0417 EN0313 ENS0319 ENS0303 EN0309 EN0207 ENS0408 EN0306 EN0408 ENS0416 EN0219 ENS0405 EN0214 EN0418 EN0308 EN0408 EN0407 ENS0205 EN0418 ENS0215 ENS0410 ENS0313 EN0303 ENS0317 EN0213 EN0415 ENS0212 ENS0413 EN0312 EN0318 ENS0418 EN0416 ENS0309 ENS0205 ENS0213 ENS0410 ENS0213 EN0220 ENS0418 ENS0219 EN0412 EN0314 ENS0419 EN0208 ENS0305 ENS0417 ENS0219 ENS0209 EN0311 ENS0412 ENS0203 ENS0417 EN0313 ENS0406 ENS0416 ENS0304 ENS0313 ENS0314 EN0312 ENS0407 ENS0310 EN0216 ENS0315 ENS0214 ENS0306 ENS0317 EN0219 ENS0303 EN0310 EN0315 ENS0204 ENS0320 EN0411 EN0419 ENS0315 EN0407 ENS0210 ENS0204 EN0405 EN0215 EN0420 EN0404 ENS0320 ENS0414 EN0220 EN0304 EN0412 EN0203 ENS0312 ENS0215 EN0218 EN0307 ENS0308 EN0320 EN0218 EN0203 ENS0220 EN0305 ENS0314 EN0316 ENS0218 ENS0311 ENS0312 EN0306 ENS0409 ENS0319 EN0413
89169-804EA 3459.8 USD InStock
89169-804 89169-926 89169-802 89169-924 CA89169-778 89169-806 89169-928 CA89169-784 CA89169-782 CA89169-788 89169-932 89169-930 CA89169-786 CA89169-780 CA22231-178 89169-936 12999-186 CA22231-176 89169-934 89169-938 12999-188 CA89169-794 89169-940 CA89169-792 89169-944 CA89169-798 CA89169-796 89169-942 CA89169-790 89169-948 89169-946 89170-000 89170-002 89170-004 89169-950 89169-712 89169-954 89169-710 89169-952 89169-716 89169-958 89169-714 89169-956 89169-718 89169-720 89169-962 89169-960 89169-724 89169-966 89169-722 89169-964 CA89169-908 CA89169-906 CA89170-000 CA89170-002 CA89170-004 CA89169-918 CA89169-912 89169-904 CA89169-910 89169-908 CA89169-916 CA89169-914 89169-906 89169-910 CA89169-920 89169-914 CA89169-924 CA89169-802 89169-912 CA89169-922 CA89169-800 89169-918 CA89169-928 CA89169-806 89169-916 CA89169-926 CA89169-804 89169-800 CA89169-930 89169-922 89169-920 CA12999-186 CA89169-934 CA12999-188 CA89169-932 CA89169-938 CA89169-936 89169-774 89169-772 89169-778 CA89169-942 CA89169-940 89169-776 89169-770 CA12999-190 CA89169-946 CA89169-944 CA89169-948 89169-786 89169-784 CA89169-710 CA89169-952 CA89169-950 89169-788 89169-782 89169-780 CA89169-714 CA89169-956 CA89169-712 22231-178 CA89169-954 22231-176 CA89169-718 CA89169-716 CA89169-958 89169-796 CA89169-960 89169-794 CA89169-722 CA89169-964 CA89169-720 CA89169-962 89169-798 89169-792 89169-790 CA89169-726 CA89169-968 CA89169-966 CA89169-724 CA89169-728 CA89169-970 CA89169-974 CA89169-732 CA89169-972 CA89169-730 CA89169-978 89169-728 CA89169-736 89169-726 89169-968 CA89169-976 CA89169-734 CA89169-738 CA89169-982 CA89169-740 89169-972 89169-730 CA89169-980 89169-970 CA89169-986 89169-976 CA89169-744 89169-734 CA89169-742 89169-732 89169-974 CA89169-984 CA89169-748 89169-738 CA89169-988 89169-978 89169-736 CA89169-746 CA89169-750 89169-984 CA89169-992 89169-742 CA89169-990 89169-740 89169-982 89169-746 CA89169-996 CA89169-754 89169-988 89169-986 CA89169-994 CA89169-752 89169-744 89169-980 12999-190 CA89169-758 89169-748 CA89169-756 CA89169-998 89169-994 89169-752 CA89169-762 89169-750 89169-992 CA89169-760 CA89169-766 89169-998 89169-756 89169-754 89169-996 CA89169-764 89169-990 89169-758 CA89169-768 89169-764 CA89169-772 CA89169-770 89169-762 89169-768 CA89169-776 CA89169-774 89169-766 89169-760
Ergomat® Nitril and Nitril Smooth Cleanroom Anti-Fatigue Floor Mats, Ergomat
Floor Mats Traction and Anti-Fatigue Floor Mats
The Ergomat® Nitril and Nitril Smooth are made from pure nitrile rubber and is designed to increase comfort while reducing trip hazards, body pain, quality issues and sick leave.

  • Choice of smooth or coned upper surface
  • Can be autoclaved (610×915 mm only/2’x3’) and features great antibacterial cleaning properties
  • Chemical exposure will not affect the state of the mat; no curling up, swelling, or expanding
  • Moulded, bevelled edges prevent curling, eliminating trip hazards
  • Raw, virgin material guarantees durability and ergonomic benefits
  • Free from silicone and latex
  • Can be pressure washed to 1,500 PSI
  • pH resistance from 5-10
  • Black color

It is ideal for areas where heat, oil and petrochemical resistance is crucial. The Nitril mats are a general application non-porous rubber ergonomic mat that is will not curl, swell or otherwise deteriorate when exposed to oils. In addition to proven ergonomic benefits, the Nitril provides exceptional traction in wet or oily environments for added worker safety. In situations with excessive fluid volume, or the need to promote air flow, Nitril can be die-cut to accommodate these requirements.

The smooth version is ideal for areas where trolleys and carts are used.

Certifications: Mats are USDA approved, meet CIFA standards for food processing, and are EN ISO Class 5(100) cleanroom rated.

Ordering information: Three-year warranty.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty