Would you like to log in again?
ERGOMAT® Complete Bubble Antifatigue Floor Mats, ERGOMAT®
Supplier: ERGOMAT INC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ERGOMAT®
STF0205 STF0305 STF0312 STF0211 STF0305 STF0306 STF0308 STF0313 STF0319 STF0415 STF0218 STF0316 STF0409 STF0219 STF0218 STF0419 STF0414 STF0217 STF0215 STF0213 STF0318 STF0309 STF0209 STF0418 STF0406 STF0408 STF0304 STF0214 STF0216 STF0307 STF0207 STF0205 STF0219 STF0206 STF0208 STF0212 STF0418 STF0210 STF0211 STF0413 STF0315 STF0417 STF0217 STF0310 STF0311 STF0405 STF0308 STF0405 STF0220 STF0309 STF0307 STF0409 STF0314 STF0417 STF0320 STF0316 STF0203 STF0304 STF0203 STF0404 STF0207 STF0411 STF0412 STF0314 STF0311 STF0208 STF0213 STF0318 STF0303 STF0310 STF0416 STF0306 STF0212 STF0412 STF0209 STF0407 STF0415 STF0410 STF0215 STF0303 STF0204 STF0313 STF0214 STF0420 STF0315 STF0210 STF0312 STF0216 STF0220 STF0416 STF0420 STF0411 STF0204 STF0414 STF0408 STF0413 STF0206 STF0419 STF0404 STF0317 STF0319 STF0407 STF0320 STF0406 STF0410 STF0317
89170-380EA 1996.35 USD InStock
89170-380 CA89170-390 89170-382 89170-384 CA89170-392 89170-386 CA89170-394 89170-420 CA89170-396 89170-388 CA89170-398 CA12999-192 89170-390 89170-392 89170-394 89170-396 CA12999-194 89170-398 CA12999-196 CA89170-322 CA89170-324 CA89170-326 CA89170-328 89170-360 89170-362 CA89170-370 CA89170-372 89170-364 CA89170-374 89170-366 CA89170-410 89170-400 CA89170-376 89170-368 CA89170-412 89170-402 CA89170-378 CA89170-414 89170-404 CA89170-416 89170-406 89170-408 CA89170-418 89170-370 CA89170-380 CA89170-382 89170-372 89170-374 CA89170-384 89170-376 CA89170-386 89170-410 89170-378 CA89170-388 89170-412 CA89170-420 89170-414 89170-416 89170-418 89170-340 CA89170-350 89170-342 89170-344 CA89170-352 89170-346 CA89170-354 CA89170-356 89170-348 CA89170-358 CA89170-360 89170-350 CA89170-362 89170-352 CA89170-364 89170-354 89170-356 CA89170-366 89170-358 CA89170-368 CA89170-400 CA89170-402 CA89170-404 CA89170-406 CA89170-408 12999-194 12999-192 12999-196 CA89170-330 89170-322 89170-324 CA89170-332 89170-326 CA89170-334 89170-328 CA89170-336 CA89170-338 89170-330 CA89170-340 CA89170-342 89170-332 89170-334 CA89170-344 CA89170-346 89170-336 CA89170-348 89170-338
ERGOMAT® Complete Bubble Antifatigue Floor Mats, ERGOMAT®
Floor Mats Traction and Anti-Fatigue Floor Mats
Ideal mat for demanding environments or where static electricity control is important.
  • Nonporous, nonslip, beveled edges, no hollow bubbles
  • Fire retardant, resistant to various oils, liquids, and acids, antistatic
  • For use in nonabrasive manufacturing environments, laboratories with occasional chemical spills, ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanrooms, or areas with little moisture
Durable gray mats are made of pure polyurethane and are designed to increase comfort while reducing tripping hazards, body aches, and other common problems in standing environments. They provide increased stimulation and blood circulation in workers' legs, reducing muscle fatigue and injury. Nonporous, nonslip mats are fire retardant; resistant to a moderate amount of oils, liquids, and acids; and have good antistatic properties. Mats are solid with no hollow bubbles and feature four molded edges that are beveled to prevent curling and tripping hazards. Suitable for use in nonabrasive manufacturing environments, laboratories with occasional chemical spills, ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanrooms, or areas with little moisture.
Ordering information: Manufacturer’s three-year warranty. Mats are available in any size or shape; contact your Avantor sales representative for more information.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty