Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Carboys with Versatile Caps, HDPE
Supplier: VWR
VWR®
152-3111-VWR 152-2211-VWR 152-1111-VWR 152-3121-VWR 152-3211-VWR 152-5221-VWR 152-7121-VWR 152-1211-VWR 152-2111-VWR 152-3221-VWR 152-5121-VWR 152-0111-VWR
89170-774EA 419.38 USD InStock
89170-774 89217-552 89170-764 89170-776 89170-766 89170-778 89170-768 89170-780 89199-164 89170-770 89170-772 89170-762
VWR® Carboys with Versatile Caps, HDPE
Bottles Carboys
VWR® HDPE carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use.
  • Chemical-resistant
  • HDPE construction
  • Molded grips for easy lifting, handling, and pouring
The distinctive rectangular shape saves valuable bench space.
VWR® HDPE carboys are molded from a translucent USP Class VI, and FDA grade resin. They offer excellent chemical resistance, making them ideal for most buffers and reagents as well as powders and solids. Versatile caps, adapters, versatile barbs, and spigots are constructed of polypropylene. The recommended autoclaving cycle for versatile caps, adapters, spigots, and versatile barbs is 20 minutes at 15 psi and 121 °C (250 °F).
Made in the USA.
Large handles on top of the container and molded handles in the base provide a full, secure grip to make lifting, carrying, and pouring easy. Specified models are available with a 120 mm wide neck, which allows the user to reach inside the carboy for easier cleaning. Leakproof polypropylene versatile caps offer the ultimate in connection flexibility. Open caps (sold separately) are compatible with interchangeable adapters, which feature a wide variety of top-connections, including molded-in hose barbs and quick-connect hose barbs. All carboys have their material and volume clearly imprinted on the outside of the bottle, as well as large, easy to read metric graduation marks which are certified to ±5% accuracy.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty