Would you like to log in again?
Aqui-Pad™ Benchtop Absorbent Mat, Therapak®
Supplier: Therapak
10324
89170-938CS 194.82 USD InStock
89170-938
Aqui-Pad™ Benchtop Absorbent Mat, Therapak®
Surface Protectors Bench Protectors
Aqui-Pad™ is specifically designed for use on the laboratory benchtop to absorb spills or drops of aqueous liquid during routine lab work
Aqui-Pad™ consists of two material layers with absorbent media sealed inside. The top layer is a non-woven polypropylene pad and the bottom layer is impervious polypropylene film to enclose any liquid spills so the liquid does not reach the benchtop. Inside the two layers is a proprietary absorbent media fabricated from tissue paper and wood pulp materials that are impregnated with a super absorbent polymer. Aqui-Pad™ is supplied in flat sheets 18" x 11" (457mm x 280mm) and will absorb up to 750 mL. Place in biohazard disposal after use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty