Would you like to log in again?
CLIA Exempt Cotinine/Nicotine Screen, Alere™ Toxicology
Supplier: Alere Toxicology
iScreen® OFD NicQuick™
DCT-102 I-DCT-B702
89171-336PK 144.54 USD InStock
89171-336 89423-388
CLIA Exempt Cotinine/Nicotine Screen, Alere™ Toxicology
Drug Tests
One-step screen tests for the qualitative detection of cotinine in human urine or saliva
The iScreen® OFD Cotinine/Nicotine Screen is a self-contained screen for the qualitative detection of cotinine in oral fluids at 30ng/mL. The screen detects 1–2 days after nicotine use. Results available in 10 minutes. The NicQuick cassette for the qualitative detection of cotinine in urine at 200ng/mL. Cassette requires only three drops of urine, and provides results in 5 minutes (pipet included).
Room temperature storage, long shelf-life. Collection Cup is not included.
Cotinine is an alkaloid found in tobaccco and is a metabolite of nicotine.
Ordering information: Collection cup is not included. The iScreen® Drug Control Cotinine Specific 2X Positive (5mL) contains positive control only. Refrigerated, must be shipped next day.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty