Would you like to log in again?
HyFlex® 11-510 Cut Protection Gloves with Plaiting on Palm and Back, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyFlex®
205743 205746 205747 205748 205745 205744
89171-386CS 2161.22 USD InStock
89171-386 89171-376 89171-384 89171-382 89171-380 89171-378
HyFlex® 11-510 Cut Protection Gloves with Plaiting on Palm and Back, Ansell
Gloves
Designed to provide cut protection in applications where abrasion resistance and dexterity are a 'must have', these gloves protect hands from abrasion in the palm area as well as the back of the hand, providing more confidence when working in restrictive spaces.
  • Excellent resistance to cuts and abrasions
  • Provide confidence in restrictive work spaces
  • Silicone-free
  • Latex-free
  • Minimal shrinking when laundered
Knitted with DuPont™ Kevlar® yarn, gloves offer flexibility and comfort along with ANSI Level 2 cut protection.
Diverse HyFlex® styles provide the best-in-class comfort, protection and dexterity required for the precise manipulation of small parts, controlled operation of power tools and gear, and direct handling of heavy parts and materials.
Use in the aerospace, automotive and transportation, construction, metal fabrication, plastic extrusion, public utilities, and white goods industries. Uses include carpentry; pipe fitting; final assembly; glass installation; maintenance operations; sheet metals and metalwork; small parts stamping; small scale metal stamping and assembly; and wiring.
The ComfortFlex cuff provides additional protection as well as a soft feel around the sensitive wrist area. Features DuPont™ Kevlar® stretch liner with spandex and patented Nylon and Kevlar® zonal plaiting on palm and back. Black foam nitrile ensures a sure grip in both light oil and dry applications. Gloves are silicone- and latex-free. Machine launderable with minimal shrinking and good recovery for use again and again.
Certifications: ANSI Level 2 cut protection; ANSI Abrasion Level 4.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags per case.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty