Would you like to log in again?
Mix-n-Stain™ CF® Dye Antibody Labeling Kits, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
92304 92237 92270 92287 92254 92246 92232 92236 92333 92262 92319 92271 92281 92255 92323 92426 92258 92275 92245 92234 92288 92322 92279 92248 92324 92338 92247 92433 92303 92230 92251 92283 92274 92252 92256 92331 92289 92291 92265 92235 92253 92238 92240 92425 92429 92431 92277 92280 92290 92320 92332 92317 92316 92250 92278 92428 92282 92242 92273 92260 92259 92241 92334 92335 92337 92336 92239 92257 92432 92243 92339 92284 92233 92264 92267 92305 92261 92268 92430 92285 92276 92231 92318 92321 92263 92272 92427
89171-584EA 159.23 USD InStock
89171-584 89171-540 89171-582 89171-580 89411-618 76404-042 76404-044 76404-046 76221-502 89171-548 89171-546 76221-504 89171-544 89171-588 89171-586 89411-620 89171-542 76221-500 89171-552 89171-596 89171-594 89171-550 89171-592 76404-050 89171-590 76404-052 76404-054 76404-056 76404-058 76404-048 89171-518 89171-516 89171-558 89171-514 89171-512 89171-556 89171-598 89171-554 89171-562 10119-160 89171-560 75832-518 75832-636 75832-830 75832-832 75832-520 89171-528 89171-606 89171-526 10119-158 89171-604 89171-524 89171-568 89171-602 89171-522 89171-566 89171-600 89171-520 89171-564 89411-608 75832-808 89171-574 89171-530 89171-572 89171-570 89411-606 75832-806 10119-156 75832-526 75832-528 75832-642 75832-522 75832-524 76221-494 89427-122 76221-496 89427-124 89427-120 89171-538 89411-612 89171-536 89411-614 76221-498 89171-578 89171-534 89171-576 89171-532
Mix-n-Stain™ CF® Dye Antibody Labeling Kits, Biotium
Protein Linking and Labeling Systems
Label your antibody with one of our superior CF® Dyes in 30 minutes without a purification step. Labeling tolerates many common buffer components including BSA and ascites.
  • Label 5 to 20 µg, 20 to 50 µg, or 50 to 100 µg Ab
  • Choice of 32 CF® Dye colors
  • Less than 30 seconds of hands-on time
  • 30 minutes total reaction time
  • No purification, 100% recovery
  • Compatible with BSA, gelatin, ascites
Mix-n-Stain™ CF® Dye Antibody Labeling Kits allow you to label ≤5 ug up to 100 ug of your antibody in just 30 minutes, with minimal hands-on time and no purification.
Mix-n-Stain™ labeling tolerates common antibody buffer formulations. An ultrafiltration centrifuge vial is provided to quickly remove interfering substances like glycerol, or to concentrate your antibody. With a slight modification in protocol, antibodies can be labeled in the presence of BSA, gelatin, or ascites fluid. Because CF® Dyes are highly water soluble, the presence of other proteins like BSA or gelatin in the labeling reaction has minimal effect on background fluorescence, because any labeled non-antibody proteins readily wash away during immunofluorescence staining.
These kits feature a choice of 32 of Biotium’s next-generation CF® Dyes, which were designed for advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Mix-n-Stain™ Antibody Labeling Kits are very simple to use (see the workflow overview). But before you begin, you must check that your antibody meets the compatibility requirements for labeling, and choose the right labeling protocol. See the product table below for a quick reference of the maximum excitation and emission of each CF® Dye.
Biotium now offer Mix-n-Stain™ Maxi Antibody Labeling Kits for labeling 1 mg of antibody with CF® Dyes, and Mix-n-Stain™ Nanobody Labeling Kits. Mix-n-Stain™ kits also are available for labeling antibodies with enzymes (HRP, AP, and GOx), fluorescent proteins (PE, APC, PerCP, and tandem dyes), biotin, or DIG.
Delivery information: Kit components: Ultrafiltration spin vial (for antibody concentration or buffer exchange, if needed), 10X Mix-n-Stain™ reaction buffer, Lyophilized Reactive CF® Dye and Mix-n-Stain™ storage buffer.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty