Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
HI-Control™ Competent Cells, Biosearch Technologies
Supplier: Biosearch Technologies
60110-1 60110-1 60435-1 60435-1
89171-614EA 410.24 USD InStock
CA89171-614 89171-614 CA89171-612 89171-612
HI-Control™ Competent Cells, Biosearch Technologies
Competent Cells
  • Tightly regulated protein expression for promoter systems using the lac operator
  • Compatible with T7-lac and other lac based promoters
  • HI-Control™ 10G Chemically Competent cells: ≥ 1×109 cfu/µg
  • HI-Control™ BL21(DE3) Chemically Competent cells: ≥1×108 cfu/µg
Competent Cells include Control DNA and Recovery Medium, and are packaged as SOLOs (1 transformation per tube). Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty