Would you like to log in again?
VWR CARRIER ACID/SOLV BOTTLE
Supplier: VWR International
Acid/Solvent Bottle Carrier^ Shatter resistant polyethylene carrier. Heavy-gauge steel wire bail has durable epoxy coating. Ribs in sides and bottom suspend filled bottle. 16mm opening allows delivery tube to be inserted. Diameter: 18cm (7.125in). Height: 35cm (13.5in). Holds 2.5L bottle.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR CARRIER ACID/SOLV BOTTLE 89171-914 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty