Would you like to log in again?
Black Dragon® Powder-Free Black Latex Examination Gloves, Microflex®, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Black Dragon®
BD-1004-PF-XL BD-1003-NPF BD-1002-PF-M BD-1001-PF-S BD-1000-NPF BD-1002-NPF BD-1000-PF-XS BD-1003-PF-L BD-1004-NPF BD-1001-NPF
89173-094BX 34.24 USD InStock
89173-094 CA89173-092 CA89173-090 89173-092 89173-088 CA89173-086 89173-086 CA89173-094 CA89173-088 89173-090
Black Dragon® Powder-Free Black Latex Examination Gloves, Microflex®, Ansell
Gloves
Black Dragon® gloves are easy to slip on, and the fully textured surface gives a better and more secure grip on tools and equipment.
  • Offer comfort and strength in a distinctive black gloves that provides better framing effect
  • Meet maximum powder and maximum water extractable protein requirements
  • Fully textured finish and beaded cuffs
  • Thickness: 5.1 mil (finger)
Combination of comfort and strength in a distinct black glove.
Fully textured for excellent tactile sensitivity. Meets maximum powder and maximum water extractable protein requirements.
Certifications: Black Dragon® gloves are manufactured under an ISO 13485 quality system and are lab and field tested to meet or exceed stringent ASTM criterion to withstand pinhole defects for unsurpassed protection and peace of mind.
Packaging: Packaged 100 per box.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty