Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
LDO and LBOD Electrodes for HQd Meter, Hach
Supplier: Hach
LDO10101 LDO10105 LDO10105 LDO10101 LBOD10101 LBOD10101
89174-014EA 2554.85 USD InStock
CA89174-018 89174-014 CA89174-014 89174-016 CA89174-016 89174-018
LDO and LBOD Electrodes for HQd Meter, Hach
Electrodes Dissolved Oxygen Electrodes
IntelliCAL LDO101 is a digital, luminescent dissolved oxygen (LDO) probe. The LDO101 is ideal for measuring dissolved oxygen in wastewater, drinking water and general water quality applications.
Hach IntelliCAL LDO is a revolutionary breakthrough in the measurement of dissolved oxygen, eliminating the numerous reliability and maintenance concerns inherent of older DO measurement technologies
IntelliCAL LBOD101 is the latest embodiment of Hach's patented Luminescent Dissolved Oxygen (LDO) technology. The LBOD101 is a digital, luminescent dissolved oxygen (LDO) sensor with a replaceable, integrated stirring system. The IntelliCAL LBOD101 probe has no membranes to foul or replace, no electrolyte solution to replenish, and fast, reliable 2-point calibration that stabilizes quickly.
The IntelliCAL LDO probe has no membranes to clean or replace, no electrolyte solution to replenish, and has reduced calibration frequency. The probes alert users when re-calibration is needed and eliminate all guesswork, providing the most reliable and accurate results with each measurement.
Certifications: .CE..
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty