Would you like to log in again?
sensION+ ORP Electrodes, Hach
Supplier: Hach
sensION+
LZW5056970002 LZW5057970002 LZW5055970002
89174-202EA 769.23 USD InStock
89174-202 89174-204 89174-226
sensION+ ORP Electrodes, Hach
Electrodes Redox Electrodes
sensION+ ORP electrodes have a mill volt range of ± 2000 mV and are equipped with a platinum wire
sensION+ ORP electrodes can be used in a variety of applications such as oxide-reduction titrations, galvanic baths, general applications, etc.
ORP probes for the sensION+ have several common features, such as a ceramic diaphragm and a glass body.
Certifications: .CE.WEEE.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty