Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Bard-Parker® Stainless Steel Scalpel Blades, Nos. 10–15, Sterile, Aspen Surgical
Supplier: Aspen Surgical
Bard-Parker®
371215 371211 371210 371212
89176-366CS 474.28 USD InStock
89176-366 89176-368 89176-362 89176-364
Bard-Parker® Stainless Steel Scalpel Blades, Nos. 10–15, Sterile, Aspen Surgical
Scalpels and Scalpel Assemblies Scalpel Blades
Stainless steel blades are sterile and individually packaged
For use with No. 3, No. 3L, No. 3LA, No. 5, No. 7, and No. 9 Bard-Parker® nonsterile surgical handles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty