Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Bard-Parker® Rib-Back™ Carbon Steel Scalpel Blades, Nos. 10–15, Non sterile, Aspen Surgical
Supplier: Aspen Surgical
Bard-Parker® Rib-Back™
371315 371310 371311 371312
89176-376CS 133.14 USD InStock
89176-376 89176-372 89176-374 89176-370
Bard-Parker® Rib-Back™ Carbon Steel Scalpel Blades, Nos. 10–15, Non sterile, Aspen Surgical
Scalpels and Scalpel Assemblies Scalpel Blades
For use with No
3, No. 5, or No. 7 Bard-Parker® scalpel handles. Fine blade carbon steel. The Rib-Back™ prevents weaving and ensures greater accuracy in the line of cut. Specially ground and carefully inspected. Individually wrapped.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty