Would you like to log in again?
SP Bel-Art Prep-Safe™ Microcentrifuge Tube Mini Floating and Vortexing Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
F187420123 F187420120 F187430127 F187420122 F187420124 F187420126 F187420125 F187420121
89176-464CS 554.5 USD InStock
89176-464 89176-466 89176-472 89176-460 89176-474 89176-462 89176-470 89176-468
SP Bel-Art Prep-Safe™ Microcentrifuge Tube Mini Floating and Vortexing Racks, Bel-Art Products, a part of SP
Racks Tube Racks
Unique tabs protect your samples from loss and contamination.
  • Polypropylene construction
  • Keep tubes in place and caps closed when under pressure
  • Autoclavable, 120 °C (250 °F)
  • Securely holds tubes in water bath, ice bucket, or benchtop
Place tubes in slots and slide under tabs to keep tubes in place and caps closed when under pressure, floating or submerged. Use in water baths, ice or on the benchtop. Fits standard 100 mL beakers and holds twelve 1.5 mL or 2.0 mL microcentrifuge tubes in numbered locations. Optional/removable vortexing attachment saves time by letting you vortex all samples simultaneously in the rack. Autoclavable polypropylene. Patent Pending.
Ordering information: All colors are available with or without removable vortexing attachment. Vortexing attachment also sold as a separate accessory.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty