Would you like to log in again?
Nunc® Carrier Plate for Cell Culture Inserts, Thermo Scientific
Supplier: Thermo Scientific
Nunc®
141082 141008 141004 141008 141006 141086 141002 141086 141078 141082 141004 141006 141078 141080 141080 141002
89177-080CS 122.87 USD InStock
89177-080 CA89177-082 89177-084 89177-072 CA89177-080 89177-082 CA89177-086 CA89177-074 89177-076 CA89177-084 89177-086 CA89177-072 89177-074 CA89177-078 89177-078 CA89177-076
Nunc® Carrier Plate for Cell Culture Inserts, Thermo Scientific
Microplates Insert Plates and Inserts
Nunc® Carrier Plates are designed for versatility in your research. The system for manipulating multiple inserts can be used for individual compartment cultures or cultures with shared media. Use with up to 12 or 24 inserts at a time.
  • Meets ANSI™ standards
  • Equipped with alphanumeric well identification
  • Polycarbonate membrane is available in three pore sizes for enhanced flexibility in your research
  • Inserts are ideal for for 3-D imaging; translucent membrane becomes transparent when wet
  • Carrier plates come in two formats that correspond with our 12- and 24-well Thermo Scientific™ Nunclon™ Delta-treated dishes
  • Uniquely engineered carrier plate features a support structure that enables variable height
  • Simply rotate the insert to engage one of the three height-adjusting tabs into the carrier plate supports
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty