Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nunclon™ Delta MultiDish Inserts, Thermo Scientific
Supplier: Thermo Scientific
Nunclon™
140640 140652 140620 140656 140663 140656 140627 140652 140629 140642 140668 140644 140627 140668 140663 140654 140660 140660 140620 140629 140654
89096-058CS 418.01 USD InStock
89096-058 11006-424 89094-366 CA89096-058 89094-368 89177-132 89177-142 CA89177-140 89177-140 CA11006-424 CA89177-134 89177-136 CA89177-144 89094-370 CA89177-132 89177-134 89177-144 CA89177-142 CA89177-138 89177-138 CA89177-136
Nunclon™ Delta MultiDish Inserts, Thermo Scientific
Culture Dishes Cell Culture Dishes
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes
Use for transport studies, toxicity tests, chemotaxis studies, and electron microscopy. They are also suitable for phase contrast and Nomarski optic systems. Inserts have been treated and tested for cell attachment, providing easy cultivation of most cell types without the need for matrix coating. They feature tabs for better handling. Nontoxic. Nonpyrogenic.
Ordering information: Packed in resealable bags.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty