Would you like to log in again?
VACUUBRAND® Vacuum Pump Oil B, BrandTech
Supplier: BrandTech
687011 687010
89178-196EA 108.12 USD InStock
89178-196 89178-198
VACUUBRAND® Vacuum Pump Oil B, BrandTech
Lubricants
Ensure the reliability of your VACUUBRAND® Rotary Vane or Chemistry-HYBRID vacuum pump by using Pump Oil B
  • Optimized for VACUUBRAND rotary vane and Chemistry-HYBRID pumps
  • Low vapor pressure, even at high temperatures
  • Excellent stability for Chemistry applications
  • 1- and 5-liter containers
Pump Oil B has excellent viscosity characteristics, good chemical resistance, low vapor pressure even at high temperatures, and better stability when pumping oxidants such as acid and alkaline vapors. Superior to standard mineral oils. Suitable for all VACUUBRAND® RE-, RZ- and RC-series pumps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty