Would you like to log in again?
Fraser Broth, Modified, HiMedia
Supplier: HiMedia Laboratories
M1764-500G
89178-272EA 271.54 USD InStock
89178-272
Fraser Broth, Modified, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
It is recommended for the selective enrichment of Listeria species from foods.
  • Based on the formulation by Fraser and Sperber
  • Contains peptic digest of animal tissue, casein enzymic hydrolysate, yeast extract and beef extract
  • Selective enrichment of Listeria species from foods
Listeria species are widely distributed and are isolated from soil, decaying vegetable matter, sewage, water, animal feed, fresh and frozen poultry, meats, raw milk, cheese and asymptomatic human and animal carriers
Fraser Broth Base, Modified, is based on the formulation by Fraser and Sperber. It is recommended for selective enrichment of Listeria species from foods.
This medium contains peptic digest of animal tissue, casein enzymic hydrolysate, yeast extract and beef extract which provide essential nutrients like carbon and nitrogenous compounds including vitamins, amino acids and trace ingredients. Phosphates buffer the medium while sodium chloride maintains osmotic equilibrium. Nalidixic acid and Acriflavin inhibits the growth of gram-negative and gram-positive organisms respectively except Listeria species. Listeria speciesc hydrolyze esculin to glucose and esculetin. The latter combines with ferric ions of ferric ammonium citrate (FD141), resulting
in the formation of 6-7 dihydroxycoumarin, a black brown complex. Ferric ammonium citrate also enhances the growth of L. monocytogenes. The high salt tolerance (of sodium chloride) of Listeria is used as means to inhibit the growth of Enterococci. Lithium chloride is also used to inhibit Enterococci, which also possess the ability to hydrolyze esculin.
Only Listeria monocytogenes from the genus Listeria ; causes infections in humans. L. monocytogenes primarily causes meningitis, encephalitis or septicemia in humans. In pregnant women, Listeria monocytogenes often causes an influenza like bacteremic illness that, if untreated, may lead to ammionitis and infection of the fetus, resulting in abortion, still birth or premature birth.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty