Would you like to log in again?
Warthin-Starry silver plating kit for histology
Supplier: AZER SCIENTIFIC INC
ES4821
89178-324EA 431.62 USD InStock
89178-324
Warthin-Starry silver plating kit for histology
Warthin-Starry silver plating kit
The Warthin-Starry Stain Kit is intended for use in the visualization of Spirochetes, Helicobacter pylori, Legionella pneumophila, and Cat Scratch Fever bacteria.
  • For Use on Formalin-Fixed, Paraffin-Embedded, or Frozen Sections
  • Histological Applications
  • Black Visualizations for All Except Klebsiella (Black/Brown), Nuclei (Brown), and the Background (Yellow)
This kit may be used on formalin-fixed, paraffin-embedded, or frozen sections.
Kit components: Spirochete sensitizer (125mL), Gelatin 4% acidulated (2 x 65mL), Silver nitrate solution 0.5% acidulated (125mL), Hydroquinone solution 0.1% acidulated (2 x 30mL), Silver nitrate solution 2% acidulated (30mL).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty