Would you like to log in again?
Versaflo™ Breathing Tubes, 3M™
Supplier: 3M
BT-20L BT-30 BT-20S 7100134625 7100009685
89178-408CS 109.76 USD InStock
89178-408 CA89178-406 89178-412 CA89178-412 89178-406
Versaflo™ Breathing Tubes, 3M™
Respirators Respiratory Systems
Light-duty breathing tubes feature steel core coils for durability.
The length adjusting breathing tube features a flexible helix that adjusts to the appropriate length for the individual wearer. The 89178-412 tube can connect any Versaflo™ headtop to any Versaflo™ air source.
All tubes feature the QRS (Quick Release Swivel) connection for maximum comfort and ease of use. Color: black with blue.
The 89178-406 and 89178-408 tubes can connect 3M™ S-Series hoods and headcovers to 3M™ powered air purifying respirators (PAPR) and supplied air respirator valves.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty