Would you like to log in again?
Axygen® Gel Cutting Tips, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
TGL-1165-R TGL-1165 TGL-1140 TGL-1140-R
89179-798CS 1128.15 USD InStock
89179-798 89179-796 89179-794 89179-792
Axygen® Gel Cutting Tips, Corning
Pipette Tips
Axygen® Gel Cutting Tips are designed to remove bands from agarose gel.
  • Designed for removing bands from agarose gel
  • One handed operation
  • Push button gel release
  • Ejector button tip release
  • Avoid cross contamination
  • Tips fit standard 1000 µL pipettors
These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty