Would you like to log in again?
Axygen® Gel Loading Tips, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
TGL-200FT37R TGL200RD57RS TGL-10FT-17R TGL-200RD57R TGL-200FT-37-R-S
89179-802CS 794.08 USD InStock
89179-802 75816-216 89179-808 89179-790 89179-806
Axygen® Gel Loading Tips, Corning
Pipette Tips
Axygen brand gel loading tips have a universal fit for reseach grade pipettors.
They have a convenient graduation mark at 5 µl for quick volume check. The 0.17 mm tips are for loading 0.2 mm thick gels.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty