Would you like to log in again?
Mobile Stainless Steel Instrument Table, Lakeside Manufacturing
Supplier: Lakeside
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8385 8382 8389 8384 8392 8399 8388 8395 8358 8356 8398 8354 8355 8359 8393 8380 8383 8394 8357 8390 8386 8391 8353 8387 8381
89182-994KT 10962.77 USD InStock
89182-994 89182-950 89182-972 89182-974 89182-952 89182-976 89182-954 89182-956 89182-978 89182-958 89182-990 89182-992 89182-970 89182-960 89182-982 89182-962 89182-984 89182-986 89182-964 89182-988 89182-966 89182-968 89182-946 89182-948 89182-980
Mobile Stainless Steel Instrument Table, Lakeside Manufacturing
Furniture Tables and Workbenches
Stainless steel instrument table.
  • All welded construction increases durability and makes surface smoother for cleaning
  • 18 gauge reinformcements supply extra security for larger models
  • Nesting tables save space
18 gauge stainless steel top (and shelf if provided) is easy to clean and sanitize. 1" square 16-gauge stainless steel legs for superior strength. 3" locking casters ensure table is stationary when needed.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty