Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
TRACK-IT™ DC Voltage and DC Current Logger, Monarch Instrument
Supplier: Monarch
TRACK-IT™
5396-0512 5396-0524 5396-0521 5396-0522 5396-0521-CAL 5396-0513 5396-0523-CAL 5396-0514 5396-0515 5396-0525-CAL 5396-0515-CAL 5396-0511 5396-0525 5396-0524-CAL 5396-0522-CAL 5396-0523 5396-0511-CAL 5396-0513-CAL 5396-0514-CAL 5396-0512-CAL
89184-560EA 464.79 USD InStock
89184-560 89184-550 89409-820 89409-810 89409-822 89409-812 89409-814 89409-804 89409-816 89409-806 89409-818 89409-808 89184-548 89184-558 89184-566 89184-556 89184-564 89184-554 89184-562 89184-552
TRACK-IT™ DC Voltage and DC Current Logger, Monarch Instrument
Data Loggers
The DC voltage and current data loggers are two channel battery powered stand alone compact data loggers that record up to 32000 samples each of DC voltage or current data
The unit is easily configured using the included Track‐It™ Software. Simply plug the logger into an open USB port on your PC and the software automatically identifies the logger. Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. After the logger has been programmed, disconnect it from the PC and attach the voltage or current module.
The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours. The on board data storage is non‐volatile so data will not be lost in the event of a depleted battery.
Units are available in 4 DC voltage ranges or for DC current. The loggers come with and LCD display with the option of a standard (1 year) or long life battery (3 year).
Certifications: CE marked.
Ordering information: Interactive software included.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty