Would you like to log in again?
VWR® Ergonomic Smooth Floor Mats, Blue
Supplier: VWR
VWR®
INS0408BLUVWR INS0320-BLU INS0307-BLU INS0309BLUVWR INS0203BLUVWR INS0204BLUVWR INS0420-BLU INS0310BLUVWR INS0305BLUVWR INS0303BLUVWR INS0412-BLU INS0304BLUVWR INS0303BLUVWR INS0210BLUVWR INS0204BLUVWR INS0209-BLU INS0206BLUVWR INS0205BLUVWR INS0410BLUVWR INS0206BLUVWR INS0203BLUVWR INS0406-BLU INS0407BLUVWR INS0208BLUVWR INS0207BLUVWR INS0220-BLU INS0405BLUVWR INS0205BLUVWR INS0308BLUVWR INS0306BLUVWR INS0304BLUVWR
89185-206EA 2232.39 USD InStock
89185-206 76318-990 89185-202 89185-126 CA89185-158 89185-128 CA89185-130 89185-122 89185-200 89185-124 89185-168 89185-162 89185-164 89185-160 76318-982 76318-984 76318-986 76318-988 CA89185-128 76318-980 89185-158 89185-136 CA89185-124 CA89185-126 89185-132 CA89185-122 89185-172 CA89185-160 76318-978 89185-196 89185-170
VWR® Ergonomic Smooth Floor Mats, Blue
Floor Mats Traction and Anti-Fatigue Floor Mats
The VWR® Smooth Mat is a true anti-fatigue mat. This smooth mat has the comfort of a polyurethane mat and the added benefit of easy clean-ability due to its closed-cell/non-porous top surface.
  • Stimulate the body to increase blood circulation and alterness
  • Molded beveled edges eliminate tripping hazards
  • Silcone free, latex free
  • Antistatic and meets ISO Class 5 (Class 100) compliance
  • Easy-to-clean, closed-cell/nonporous top surface
  • Highly resistant to chemicals and UV light
Safety features include molded beveled edges that will never curl up, eliminating tripping hazards. With the VWR® Smooth mat, you will never have overlapping or worn-out connecting mats, also eliminating tripping hazards.
Constructed of the highest quality using only virgin raw material, this mat has lifetime ergonomic benefits. The VWR® Smooth mat is silicone & latex-free. Using its unique ergonomic designs, this smooth mat prevents common problems encountered in standing environments while stimulating the body to increase blood circulation and alertness that will reduce sick leave, workers’ compensation and quality issues. The VWR® Smooth mat is a solid mat throughout.
It also can be customized to any size and any shape, always in one piece.
Certifications: Meets Cleanroom ISO Class 5 (Class 100) compliance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty