Would you like to log in again?
VWR® Ergonomic Economy Smooth Floor Mats, Black
Supplier: VWR
SB0204VWR SB0404VWR SB0206VWR SB0205VWR SB0306VWR SB0208VWR SB0207VWR SB0209VWR SB0305VWR SB0307VWR SB0305VWR SB0203VWR SB0308VWR SB0407VWR SB0308VWR SB0408VWR SB0208VWR SB0207VWR SB0410VWR SB0405VWR SB0306VWR SB0203VWR SB0309VWR SB0303VWR SB0206VWR SB0204VWR SB0310VWR SB0304VWR SB0304VWR SB0406VWR SB0320VWR
89185-448EA 290.97 USD InStock
89185-448 89185-444 89185-488 89185-446 89185-524 CA89185-478 89185-484 89185-440 CA89185-450 89185-442 89185-486 89185-520 89185-480 89185-482 89185-518 89185-514 CA89185-446 CA89185-448 89185-516 CA89185-486 CA89185-442 89185-510 89185-478 89185-512 CA89185-482 89185-452 CA89185-440 89185-476 CA89185-480 89185-450 89185-490
VWR® Ergonomic Economy Smooth Floor Mats, Black
Floor Mats Traction and Anti-Fatigue Floor Mats
The VWR® Economy Smooth Mat is a low-density polyurethane material with an anti-fatigue profile. This flat mat is ideal for dry, non-critical environments combining both sitting and standing work areas. No smooth matting available delivers as much relief and comfort at this price. The VWR® Economy Smooth Mat can be customized to any size and any shape, always in one piece.
  • Stimulate the body to increase blood circulation and alertness
  • Molded beveled edges eliminate tripping hazards
  • Silcone free, latex free
  • Easy-to-clean, closed-cell/nonporous top surface
  • Highly resistant to chemicals and UV light
Safety features include molded beveled edges that will never curl up, eliminating tripping hazards
The VWR® Economy Smooth Mat has lifetime ergonomic benefits. Using its unique ergonomic designs, this flat mat prevents common problems encountered in standing environments while stimulating the body to increase blood circulation and alertness that will reduce sick leave, workers' compensation and quality issues. The VWR® Economy Smooth Mat is a solid mat throughout. This mat is silicone & latex-free.
With the VWR® Economy Smooth Mat, you will never have overlapping or worn-out connecting mats, which also eliminates tripping hazards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty