Would you like to log in again?
Whatman™ Glass Extraction Thimble, Grade 603G, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10371011 10371042 10371036 10371114 10371007 10371114 10371025 10371043 2811-308 10371045 10371045 10371025 10371036 10371042 10371007 10371019 10371005 10371103 10371055 10371055 2811-308 10371019 10371005 10371011 10371103 10371043
10034-860PK 281.2 USD InStock
CA11013-734 10034-860 CA89187-126 CA89187-128 CA11013-570 10034-866 CA89187-122 10034-864 10034-862 CA89187-124 CA13500-142 10034-858 10035-416 CA11013-568 89187-128 CA11013-864 CA11013-566 CA11013-880 10034-856 89187-122 89187-124 89187-126 11008-576 CA11008-576 13500-142 CA11013-878
Whatman™ Glass Extraction Thimble, Grade 603G, Whatman products (Cytiva)
Extractors Extraction Thimbles
Cytiva's Whatman 603G borosilicate glass fiber thimbles with inorganic binder are flexibly designed for use in a full range of commercially available Soxhlet apparatus systems.
  • Resistant to temperatures up to 500°C (932°F)
  • Manufactured with walls as thin as 1 to 1.5 mm
  • Analysis of hot and acidic gases, including emission control and smoke stack gas monitoring
  • Soxhlet extraction
  • Analysis of pesticide residues
  • Determination of oil/fat content of foods
  • Analysis of oil and grease in solid wastes
Whatman standard glass extraction thimbles are available in a range of dimensions and wall thicknesses to suit your application.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty