Would you like to log in again?
Whatman™ Quartz Extraction Thimble for Emission Testing, Tapered, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
2812-259 2812-259
89187-130PK 144.33 USD InStock
89187-130 CA89187-130
Whatman™ Quartz Extraction Thimble for Emission Testing, Tapered, Whatman products (Cytiva)
Extractors Extraction Thimbles
High-purity Whatman quartz thimbles from Cytiva support accurate, reproducible emission testing by providing low extractable levels and temperature resistance up to 800 °C.
  • Appropriate for manual sampling in aggressive environments
  • Suitable for both solvent extraction and air sampling applications
  • High-purity quartz/silica, for high-temperature resistance (up to 800 °C)
  • No binders or chemical additives
  • Low heavy metal content
  • High collection efficiency
Each Whatman quartz thimble from Cytiva is made from high-purity quartz with low heavy metal concentrations. Thimbles are 90 mm in length, tapered for stack sample loading, and have an outside diameter of 25 mm.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty