Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heracell i Copper CO₂ Incubators, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Heracell i
51026419 51026332 51026534 51026332 50116050 51026283 51026283 50116050 51026419 51026534
89187-170EA 14032.27 USD InStock
CA97015-616 CA97015-614 CA97015-648 89187-170 89187-182 89187-174 89187-186 CA97015-650 89187-178 CA97015-618
Heracell i Copper CO₂ Incubators, Thermo Scientific
Incubators CO₂ Incubators
Thermo Scientific Heracell i CO₂ incubators provide superior security for your important cell cultures, featuring 100% pure copper interior for 24/7 protection against contaminants potentially introduced through door openings or sample handling, and the on-demand ContraCon 90° moist heat decontamination cycle that ensures worry-free cleaning and operation

Available in two conveniently stackable sizes (150 L and 240 L), Heracell incubators may be purchased with a choice of TC or IR CO₂ sensors. Thermal conductivity (TC) sensors feature accurate gas sensor technology and are known for their long service life. Infrared (IR) sensor features exclusive dual-beam technology for precise CO₂ control in fluctuating temperature and humidity conditions.

Thermo Scientific Heracell i CO₂ incubators combines optimal culturing conditions with simplicity and ease of use. In-chamber sensors and probes with an integrated, heated humidity water reservoir facilitate fast recovery rates of all parameters after routine access. The exclusive iCAN touch screen controller provides complete data visibility to monitor all incubator interactions with on-screen menu prompts, error and usage logs, data logging and trend graphing, and multiple language selection.

Independent research highlights that 100% copper eliminates 99.9% of microbial contaminants quickly and effectively. Copper’s continuous, active decontamination improves with its natural oxidation and does not disrupt or harm the cultures.

Certifications: CE marked, CSA approved.

Ordering information: Includes 100% solid copper interior, ContraCon decontamination, and iCAN touchscreen.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

SERVICES


Services: Thermo Scientific provides several service options such as Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Heracell CO2 Incubator Technical Services" in the website search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST@vwr.com for further details.

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty