Would you like to log in again?
WYPALL® X90 Cloths, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
WYPALL®
12890 12890 12889 12889 12891 12891
89194-382CS 148.86 USD InStock
CA89194-386 89194-382 CA89194-384 89194-386 CA89194-382 89194-384
WYPALL® X90 Cloths, KIMBERLY-CLARK PROFESSIONAL®
Wipes Reusable Cleaning Cloths Cloths for General Applications
The new WYPALL® X90 Cloths provide 75% more oil absorption and 35% more water absorption.
Cloths are also reusable, ensuring more use out of every sheet. They cut waste disposal, freight, and storage costs associated with shop towels and rags. They also help reduce worker exposure to harmful contaminants. Super-soft cloths are ideal for heavy wiping, prepping surfaces with solvents, and cleaning metal shavings and rough surfaces.
Cloths are manufactured by combining hydro-entangling polyester fiber for softness and oil absorbency and wood fiber for water absorbency with mega-strong spunbond material using our HYDROKNIT® Technology. The result is one half of an X90 cloth. By bonding two plies together, the new WYPALL® X90 Cloth becomes two plies of purely efficient performance. Because the cloths are more absorbent than comparable industrial wiping solutions - especially shop towels and rags - fewer cloths are required for every cleanup, every workday. Five Jumbo Rolls can absorb a 55-gallon drum of oil.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty